ContactUsLinkComponent
BDH DOUBLZONE PH PAPER1.0-12.0
pH Indicator Paper DoubleZone, VWR Chemicals BDH®
# de catalogue BDH35300.606
Fournisseur:  BDH
BDH DOUBLZONE PH PAPER1.0-12.0
pH Indicator Paper DoubleZone, VWR Chemicals BDH®
# de catalogue BDH35300.606
Fournisseur:  BDH
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • pH Range:
    1.0 - 12.0
  • pH Graduation:
    1.0
  • Supplier No.:
    90301.2047
Spécifications
A propos de cet article
These indicator papers show two different colors for each pH value at intervals of 0.3–1 pH unit, increasing the accuracy of readings and allowing good estimation of intermediate values
Paper comes on a plastic reel that will protect from many environmental influences, ensuring that the paper is always ready to use. Width: 10mm. Length: 5m.
Customers Who Bought This Also Bought