Spécifications
- pH Range:1.0 - 12.0
- pH Graduation:1.0
- Supplier No.:90301.2047
Spécifications
A propos de cet article
These indicator papers show two different colors for each pH value at intervals of 0.3–1 pH unit, increasing the accuracy of readings and allowing good estimation of intermediate values
Paper comes on a plastic reel that will protect from many environmental influences, ensuring that the paper is always ready to use. Width: 10mm. Length: 5m.