Order Entry
ContactUsLinkComponent
VWR* Oven Thermometer Bi-Metallic
VWR® Bi-Metallic Oven Thermometer
# de catalogue 98000-214
Fournisseur:  VWR International
VWR* Oven Thermometer Bi-Metallic
VWR® Bi-Metallic Oven Thermometer
# de catalogue 98000-214
Fournisseur:  VWR International

Spécifications

  • Accuracy:
    10 °C/°F
  • Ø:
    6.4 cm (21/2")
  • Cat. no.:
    98000-214
  • Divisions:
    10
  • Range:
    10...260 °C (50...500 °F)

Spécifications

A propos de cet article

Thermometer features stainless steel construction with plastic dial face.

  • Essential to ensure proper food preparation

Reliable indicator of oven temperature and "doneness" of cooked food.

Customers Who Bought This Also Bought