Specifications
- Description:pH Strips, 5.5-8.0
- Cat. no.:97058-562
- Supplier No.:9700
Specifications
About this item
These instant-read pH strips provide a simple, single color match with the largest matching area available
Packed in sturdy flip-top vials. Each vial contains 100 test strips.
Match a single pad to the color chart for a clear pH reading at each whole unit