Spécifications
- Description:pH Strips, 0.0-14.0
- Cat. no.:97058-560
- Supplier No.:9800
Spécifications
A propos de cet article
These instant-read pH strips provide a simple, single color match with the largest matching area available
Packed in sturdy flip-top vials. Each vial contains 100 test strips.
Match a single pad to the color chart for a clear pH reading at each whole unit