ContactUsLinkComponent
Ph Test Strip
Hydrion® pH strips, Micro Essential Laboratory
# de catalogue 97058-560
Fournisseur:  Micro Essential
Ph Test Strip
Hydrion® pH strips, Micro Essential Laboratory
# de catalogue 97058-560
Fournisseur:  Micro Essential
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    pH Strips, 0.0-14.0
  • Cat. no.:
    97058-560
  • Supplier No.:
    9800
Spécifications
A propos de cet article
These instant-read pH strips provide a simple, single color match with the largest matching area available
Packed in sturdy flip-top vials. Each vial contains 100 test strips.
Match a single pad to the color chart for a clear pH reading at each whole unit
Customers Who Bought This Also Bought