Spécifications
- Description:Non-Latex Tourniquet, 0.6 mm thick
- Cat. no.:97001-006
- Packaging:25 Perforated Straps/Reel
- Supplier No.:10204
Spécifications
A propos de cet article
Designed for disposable use in hospitals and clinics, these tear-resistant and slip-resistant latex straps are easily tied and untied, and can be rolled and banded
Tourniquets are available in low-powder latex or in powder-free non-latex for patients with latex sensitivities. A variety of packaging options are available. Individually precut to measure 2.5Wx45.7Lcm (1x18")