Order Entry
ContactUsLinkComponent
Tip Gel Cut 1.1 X 6.5Mm Pk250
Axygen® Gel Cutting Tips, Corning
# de catalogue 89179-796
Fournisseur:  Corning
Product available on GSA Advantage®
Tip Gel Cut 1.1 X 6.5Mm Pk250
Axygen® Gel Cutting Tips, Corning
# de catalogue 89179-796
Fournisseur:  Corning
Product available on GSA Advantage®

Spécifications

  • Length:
    11 mm
  • Tip Color:
    Clear
  • Packed:
    Bulk Pack, 250 Tips/Pack
  • Cat. no.:
    89179-796
  • Tip Style:
    Gel cutting, 6.5 mm
  • Supplier No.:
    TGL-1165

Spécifications

A propos de cet article

Axygen® Gel Cutting Tips are designed to remove bands from agarose gel.

  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 µL pipettors

These polypropylene tips are nonpyrogenic and RNase-/DNase-free.

Customers Who Bought This Also Bought