Axygen® Pipette Tips for Hamamatsu 6000 and 7000 systems, Corning
# de catalogue 89177-042
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:50 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Tip Color:Black
- Supplier No.:FDSS1536BKR
Spécifications
A propos de cet article
Axygen FDSS series tips for Hamamatsu 6000 and 7000 systems.
Axygen robotic tips are engineered to exact dimensions of OEM tips to ensure automation compatibility.
Customers Who Bought This Also Bought