VWR® Hazmat Spill Response Kit
# de catalogue 89168-998
Fournisseur: VWR International
Spécifications
- Cat. no.:89168-998
- Absorbency:34.1 L (9 gal.) of acids, caustics, bases
Spécifications
A propos de cet article
Hazmat spill response kit includes 20 pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie.
- Includes 20 pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie
- Supports compliance with 40 CFR 263.30(a) safety standard
Customers Who Bought This Also Bought