Spécifications
- Volume:1250 µl
- Environmentally Preferable:Sustainable Packaging,Safer Human and Environmental Health
- Packed:96/Rack
- Tip Style:Extended Length Tips, Universal Fit, Low Retention
- Supplier No.:BT1250
Spécifications
A propos de cet article
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency.
Eco-friendly empty pipet tip trays are manufactured from recycled polypropylene. The trays are autoclavable, enabling reuse for maximum sustainability. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
- Trays are autoclavable
- Enabling reuse for maximum sustainability
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency.
Eco-friendly empty pipette tip trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
Thin wall design provides an optimal seal for single and multichannel pipettors. Tips featuring the S³ polymer eliminate sample hold up and ensure a more precise sample draw and dispense. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration.