Neptune® BT Barrier Pipette Tips, Pre-Sterile
Catalog # 89168-750
Supplier: Biotix
Product available on GSA Advantage®
Specifications
- Volume:10 µl
- Environmentally Preferable:Sustainable Packaging,Safer Human and Environmental Health
- Packed:96/Rack
- Cat. no.:89168-750
- Tip Style:Extended Length Tips, Universal Fit, Low Retention
- Supplier No.:BT10XLS3
Specifications
About this item
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency.
- Trays are autoclavable
- Enabling reuse for maximum sustainability
Eco-friendly empty pipette tip trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
Thin wall design provides an optimal seal for single and multichannel pipettors. Tips featuring the S³ polymer eliminate sample hold up and ensure a more precise sample draw and dispense. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Customers Who Bought This Also Bought