ContactUsLinkComponent
Barrier Pipet Tip
Neptune® BT Barrier Pipette Tips, Pre-Sterile
# de catalogue 89140-932
Fournisseur:  Biotix
   Sustainable Packaging
   Safer Human and Environmental Health
Product available on GSA Advantage®
Barrier Pipet Tip
Neptune® BT Barrier Pipette Tips, Pre-Sterile
# de catalogue 89140-932
Fournisseur:  Biotix
   Sustainable Packaging
   Safer Human and Environmental Health
Product available on GSA Advantage®
Spécifications
  • Volume:
    20 µl
  • Environmentally Preferable:
    Sustainable Packaging,
    Safer Human and Environmental Health
  • Packed:
    96/Rack
  • Tip Style:
    Universal Fit Tips, Low Retention
  • Supplier No.:
    BT20
Spécifications
A propos de cet article
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency.
Eco-friendly empty pipet tip trays are manufactured from recycled polypropylene. The trays are autoclavable, enabling reuse for maximum sustainability. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
  • Trays are autoclavable
  • Enabling reuse for maximum sustainability
Pre-sterile universal barrier tips are made from virgin polypropylene and feature high density polyethylene filters to ensure against aerosol contamination and provide superior consistency.
Eco-friendly empty pipette tip trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
Thin wall design provides an optimal seal for single and multichannel pipettors. Tips featuring the S³ polymer eliminate sample hold up and ensure a more precise sample draw and dispense. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Customers Who Bought This Also Bought