ContactUsLinkComponent
1000ul Graduated Slimline Pipet Tips
Neptune® Universal Pipette Tips
# de catalogue 89140-918
Fournisseur:  Biotix
Product available on GSA Advantage®
1000ul Graduated Slimline Pipet Tips
Neptune® Universal Pipette Tips
# de catalogue 89140-918
Fournisseur:  Biotix
Product available on GSA Advantage®
Spécifications
  • Volume:
    1000 µl
  • Tip Color:
    Clear
  • Packed:
    Bulk
  • Tip Style:
    Universal Fit Tips
  • Supplier No.:
    2160
Spécifications
A propos de cet article
These universal pipette tips are made from virgin polypropylene.
Eco-friendly empty pipet tip trays are manufactured from recycled polypropylene. The trays are autoclavable, enabling reuse for maximum sustainability. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%. The trays provide superior stability, firm placement, and a maximum space saving design.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
These universal pipette tips are made from virgin polypropylene.
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
The thin wall design ensures an optimal seal and easy ejection with industry leading pipettors. Eco-friendly racks are made from recycled polypropylene and feature a unique compact packaging configuration that reduces the carbon footprint.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Ordering information: Bulk orders are packaged 1000 tips per bag. Rack & Stack packaging contains five inserts of 192 tips.
Customers Who Bought This Also Bought