Neptune® S³ Low Retention Pipette Tips
Catalog # 89140-100
Supplier: Biotix
Specifications
- Volume:10 µl
- Environmentally Preferable:Sustainable Packaging,Safer Human and Environmental Health
- Tip Color:Clear
- Packed:96/Rack
- Cat. no.:89140-100
- Tip Style:Micro Tips, Low Retention
- Supplier No.:2042
Specifications
About this item
The exclusive S³ polymer delivers to within 0.1% of indicated volume, eliminating sample hold-up and ensuring a more precise application.
- Autoclavable
- Good sustainability
- Superior stability, firm placement
- Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results. Universal pipette tips are made from virgin polypropylene and are equipped with a thin wall design that provides an optimal seal with industry leading pipettors.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
Ordering information: A variety of packaging options are available. Bulk orders are packaged 1000 per bag. Rack and Stack packaging contains five inserts of 192 tips. Standard rack packaging contains 96 tips per rack.
Customers Who Bought This Also Bought