Extra-Safe™ Knee Length Labcoats, ValuMax®
# de catalogue 89140-074
Fournisseur: Valumax International, Inc.
Spécifications
- Clothing Size:M
- Garment application:Laboratory
- Color:Pink
- Description:Extra-Safe™ Knee Length Labcoats with 3 Pockets
- Cat. no.:89140-074
- Supplier No.:3660LPM
Spécifications
A propos de cet article
These disposable lab coats are made from soft, three-layer SMS fabrics
These disposable lab coats are made from soft, three-layer SMS fabrics
They are fluid-resistant and breathable. Coats feature snap fronts, three pockets (unless otherwise noted), and knit collars. All coats are autoclavable and washable up to five times.
Ordering information: White lab coats are also available with traditional collar. Contact your Avantor sales representative for additional information.
Customers Who Bought This Also Bought