ContactUsLinkComponent
Extra-Safe Lab coat
Extra-Safe™ Knee Length Labcoats, ValuMax®
# de catalogue 89140-074
Extra-Safe™ Knee Length Labcoats, ValuMax®
# de catalogue 89140-074
Spécifications
  • Clothing Size:
    M
  • Garment application:
    Laboratory
  • Color:
    Pink
  • Description:
    Extra-Safe™ Knee Length Labcoats with 3 Pockets
  • Cat. no.:
    89140-074
  • Supplier No.:
    3660LPM
Spécifications
A propos de cet article
These disposable lab coats are made from soft, three-layer SMS fabrics
These disposable lab coats are made from soft, three-layer SMS fabrics
They are fluid-resistant and breathable. Coats feature snap fronts, three pockets (unless otherwise noted), and knit collars. All coats are autoclavable and washable up to five times.
Ordering information: White lab coats are also available with traditional collar. Contact your Avantor sales representative for additional information.
Customers Who Bought This Also Bought