Order Entry
ContactUsLinkComponent
50mL Freezer Box
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
# de catalogue 89128-210
Fournisseur:  Argos Technologies
Small Business Enterprise
50mL Freezer Box
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
# de catalogue 89128-210
Fournisseur:  Argos Technologies
Small Business Enterprise

Spécifications

  • Color:
    Orange
  • Description:
    50ml Polypropylene Storage Box
  • Cat. no.:
    89128-210
  • Supplier No.:
    R0050

Spécifications

A propos de cet article

Polypropylene freezer boxes accommodate 15 & 50mL centrifuge tubes for long term storage; an excellent alternative to cardboard storage boxes

Boxes can be stored to -90°C (130°F) and are autoclavable to 122°C (251.6°F). Accommodate most 4" stainless steel freezer racks.

Clear lid and easy-to-read numerical grid allows for tube reference and identification. White top portion allows for easy labeling or marking.

Customers Who Bought This Also Bought