Axygen® Disposable Reagent Reservoirs, Corning
Catalog # 89108-008
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Capacity:50 ml
- Description:Disposable Reagent Reservoir
- Material:Plastic
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Packed:40 Packs of 5
- Cat. no.:89108-008
- Supplier No.:RES-V-50-S
Specifications
About this item
These disposable reagent reservoirs are sterile and certified nonpyrogenic
They feature a sharp-V bottom that allows efficient extraction of the reagent. They also feature four pour-off corners and a stacking lug for easy separation
Customers Who Bought This Also Bought