VWR® Sleeve Stoppers
# de catalogue 89097-558
Fournisseur: VWR International
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Color:White
- Size:24/40
- Cat. no.:89097-558
- I.D.×O.D.:24×30 mm
Spécifications
A propos de cet article
Natural rubber stoppers feature a hollow plug to fit inside bottles, flasks, vials, or tubing.
- Natural rubber construction
- Excellent resealing properties
- Hollow plug
- Suitable for air-sensitive chemistry
Attached sleeves pull over neck of bottle to form a double seal. The stopper allows insertion of a syringe needle. Stoppers exhibit excellent resealing properties for air-sensitive chemistry.
Customers Who Bought This Also Bought