ContactUsLinkComponent
VWR STOPPER WHITE 14/20 PK100
VWR® Sleeve Stoppers
Catalog # 89097-554
Supplier:  VWR International
Product available on GSA Advantage®
Small Business Enterprise
VWR STOPPER WHITE 14/20 PK100
VWR® Sleeve Stoppers
Catalog # 89097-554
Supplier:  VWR International
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Color:
    White
  • Size:
    14/20
  • Cat. no.:
    89097-554
  • I.D.×O.D.:
    13×20 mm
Specifications
About this item
Natural rubber stoppers feature a hollow plug to fit inside bottles, flasks, vials, or tubing.
  • Natural rubber construction
  • Excellent resealing properties
  • Hollow plug
  • Suitable for air-sensitive chemistry
Attached sleeves pull over neck of bottle to form a double seal. The stopper allows insertion of a syringe needle. Stoppers exhibit excellent resealing properties for air-sensitive chemistry.
Customers Who Bought This Also Bought