ContactUsLinkComponent
250ul Violet Tips for V-Prep*
Axygen® Automation Tips for Agilent®/Velocity11 VPrep® and Bravo Workstations, Corning
# de catalogue 89096-078
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Axygen® Automation Tips for Agilent®/Velocity11 VPrep® and Bravo Workstations, Corning
# de catalogue 89096-078
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Spécifications
  • Volume:
    250 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Tip Color:
    Clear
  • Packed:
    96 Tips/Rack, 10 Racks/Pack, 5 Packs/case
  • Sterile:
    +
  • Tip Style:
    Standard
  • For Use With:
    96LT / 8LT
  • Supplier No.:
    VT-250-R-S
Spécifications
A propos de cet article
Axygen® automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Maxymum Recovery® version available in most tip styles
  • Filtered version available in most tip styles to prevent aerosol and liquid contamination
Axygen® offers a comprehensive line of tips for Agilent®/Velocity11 automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.Available in nonsterile, sterile, non-filtered, filtered, and wide-bore formats. Also available in Maxymum Recovery® to further improve accuracy and precision while reducing sample loss. Clear polypropylene tips are available in 96-well and 384-well format designed for Agilent®/Velocity VPrep® and Bravo 96LT, 8LT, 384ST, 96ST and 16ST heads.
Certifications: Tips are certified RNase-/DNase-free, and nonpyrogenic.
Customers Who Bought This Also Bought