SP Bel-Art Wash Bottles, 4-Color, Right-to-Know, Safety-Vented and Safety-Labeled, Wide Mouth, Bel-Art Products, a part of SP
# de catalogue 89094-886
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Capacity:250 ml (8.5 oz.)
- Description:Isopropanol
- Cat. no.:89094-886
- Cap Color:Yellow
- Supplier No.:F118080008
Spécifications
A propos de cet article
A unique valve prevents pressure build-up inside these Right-to-Know Safety-Labeled Wash Bottles, eliminating potentially dangerous drips.
- Wide mouth for easy refilling
- Leakproof
- Translucent LDPE construction
- Color-coded polypropylene closures for quick identification
This valve allows dispensing in an upright or inverted position.
Bottles feature a 53 mm (2") wide mouth for easy refilling. They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Leak-proof, these translucent* LDPE bottles have color-coded polypropylene closures to aid in quick and easy identification.
*Note: Use LYOB only with solvents compatible with LDPE.
Certifications: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Ordering information: The 250 and 500 ml bottles are packaged four per bag. The 1000 ml bottles are packaged two per bag. The Assortment item includes six per bag, with one each of Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and Label Your Own Bottle (LYOB).
Customers Who Bought This Also Bought