ContactUsLinkComponent
HOLDER CON. TUBE 15 & 50ML PK4
SP Bel-Art Conical Tube Holder, 15 and 50 ml, Bel-Art Products, a part of SP
# de catalogue 89093-448
HOLDER CON. TUBE 15 & 50ML PK4
SP Bel-Art Conical Tube Holder, 15 and 50 ml, Bel-Art Products, a part of SP
# de catalogue 89093-448
Spécifications
  • Description:
    Conical Tube Holder
  • Height:
    6.4 cm (2.5") Arms
  • Cat. no.:
    89093-448
  • For Tubes:
    15/50 ml
  • Supplier No.:
    187960000
Spécifications
A propos de cet article
The most versatile tube holder on the market, this set-up takes just seconds and gives users the choice of creating a 15 mL or a 50 mL tube holder.
  • Easy set-up
  • Open uprights allow clear visibility
  • Conical bottom
  • Easy to remove from holder
  • Polypropylene construction
  • Autoclavable
Unlike other tube holders, the open uprights allow users to view the entire contents of the tube
Tubes can be easily lifted from the holder, or pinch the uprights to carry the holder with the tube. Tube holders are manufactured from autoclavable polypropylene.
No more guessing if there is liquid remaining in the conical bottom.
Customers Who Bought This Also Bought