ContactUsLinkComponent
Scoop Non-Sparking
Non-Sparking Scoop, New Pig
# de catalogue 89093-176
Fournisseur:  New Pig
Scoop Non-Sparking
Non-Sparking Scoop, New Pig
# de catalogue 89093-176
Fournisseur:  New Pig
Spécifications
  • Cat. no.:
    89093-176
  • Supplier No.:
    TLS290
Spécifications
A propos de cet article
Excellent chemical resistance for use in cleaning up loose absorbents
Non-static and non-sparking construction is excellent for use with flammables. Tough, resilient polypropylene won't rust or corrode
Customers Who Bought This Also Bought