Non-Sparking Scoop, New Pig
# de catalogue 89093-176
Fournisseur: New Pig
Spécifications
- Cat. no.:89093-176
- Supplier No.:TLS290
Spécifications
A propos de cet article
Excellent chemical resistance for use in cleaning up loose absorbents
Non-static and non-sparking construction is excellent for use with flammables. Tough, resilient polypropylene won't rust or corrode
Customers Who Bought This Also Bought