JACKSON SAFETY® V20 VISIO™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
# de catalogue 89092-770
Fournisseur: Kimberly-Clark
Spécifications
- Lens Coating:Anti-fog
- Lens Tint:Smoke
- Description:V20 VISIO™ safety glasses
- Frame Color:Silver
- Cat. no.:89092-770
- Supplier No.:14473
Spécifications
A propos de cet article
Lightweight eight base curved lens provides wraparound protection for a wide field of vision.
- Unique duo-tone frame
- Universal nose bridge
Three-point adjustable temples ensure a comfortable fit. Lens provides 99.9% U.V. protection.
Certifications: Meets ANSI Z87.1+ standards.
Customers Who Bought This Also Bought