Order Entry
ContactUsLinkComponent
FLASK 1000ML 5340 6/PK
PYREX® Filtering Flasks, Heavy Wall with Tubulation, Corning
# de catalogue 89090-868
Fournisseur:  Corning
FLASK 1000ML 5340 6/PK
PYREX® Filtering Flasks, Heavy Wall with Tubulation, Corning
# de catalogue 89090-868
Fournisseur:  Corning

Spécifications

  • Capacity:
    1 L
  • Cat. no.:
    89090-868
  • Rubber Stopper Size:
    8
  • Tubulation O.D.:
    10 mm
  • Supplier No.:
    5340-1L

Spécifications

A propos de cet article

PYREX® borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.

The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.

Customers Who Bought This Also Bought