PYREX® Liebig Condensers, with Drip Tip and [ST] Outer and Inner Joints, Corning
# de catalogue 89090-726
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Description:PYREX® 300mm Liebig Condensers, with Drip Tip and 24/40 [ST] Outer and Inner Joints
- Environmentally Preferable:Low Manufacturing Impact
- Cat. no.:89090-726
- Supplier No.:2400-300
- Environmental certification:I-RECs, ISO 14001 - Monterrey
Spécifications
A propos de cet article
Made of PYREX® borosilicate glass.
- With outer joint at the top and inner standard taper joint with drip tip
- Very good temperature resistance
- Very good chemical resistance
This general purpose condenser can be used for distillation, refluxing and extraction operations. The condenser tubulations are approximately 10 mm Ext.Ø. Can be used for vacuum distillation.
Customers Who Bought This Also Bought