Order Entry
ContactUsLinkComponent
FLASK VOL PHOSPH ACID 200 ML
PYREX® Volumetric Flasks, Phosphoric Acid, Wide Neck, Corning
# de catalogue 89090-684
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
FLASK VOL PHOSPH ACID 200 ML
PYREX® Volumetric Flasks, Phosphoric Acid, Wide Neck, Corning
# de catalogue 89090-684
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®

Spécifications

  • Capacity:
    200 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    89090-684
  • Tolerance:
    ±0.18 ml
  • Supplier No.:
    5840-200

Spécifications

A propos de cet article

PYREX® borosilicate glass.

  • Calibrated 'to contain'
  • Heavily constructed throughout

For use in the determination of phosphoric acid in mixed fertilizer and more.

Customers Who Bought This Also Bought