PYREX® Erlenmeyer Flasks, Graduated, Narrow Mouth, [ST] Joint, Corning
# de catalogue 89090-646
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:125 ml
- ST Joint No.:24/40
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:89090-646
- Supplier No.:5000-125
Spécifications
A propos de cet article
PYREX® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The uniform wall thickness of these flasks provide the proper balance between mechanical strength and thermal shock resistance. They are graduated to show approximate capacity and have an extra large marking space.
Certifications: ASTM E-123
Customers Who Bought This Also Bought