Order Entry
ContactUsLinkComponent
FLASK FLTR CONNECTN 125 ML PK6
PYREX® Filtering Flasks, Heavy Wall with Tubulation, Corning
Catalog # 89090-576
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
FLASK FLTR CONNECTN 125 ML PK6
PYREX® Filtering Flasks, Heavy Wall with Tubulation, Corning
Catalog # 89090-576
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact

Specifications

  • Capacity:
    125 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    89090-576
  • Rubber Stopper Size:
    5
  • Tubulation O.D.:
    10 mm
  • Supplier No.:
    5360-125

Specifications

About this item

PYREX® borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.

The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.

Customers Who Bought This Also Bought