ContactUsLinkComponent
DISH 80X40MM 3140 6/PK
PYREX® Crystallizing Dishes, Corning
# de catalogue 89090-478
Fournisseur:  Corning
Product available on GSA Advantage®
PYREX® Crystallizing Dishes, Corning
# de catalogue 89090-478
Fournisseur:  Corning
Product available on GSA Advantage®
Spécifications
  • Cat. no.:
    89090-478
  • Diameter ×H:
    80 x 40 mm
  • Supplier No.:
    3140-80
Spécifications
A propos de cet article
PYREX® borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.
Customers Who Bought This Also Bought