PYREX® Crystallizing Dishes, Corning
# de catalogue 89090-478
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Cat. no.:89090-478
- Diameter ×H:80 x 40 mm
- Supplier No.:3140-80
Spécifications
A propos de cet article
PYREX® borosilicate glass, without spout.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.
Customers Who Bought This Also Bought