PYREX® Powder Funnels, Glass, Corning
Catalog # 89090-472
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Environmentally Preferable:Low Manufacturing Impact
- Cat. no.:89090-472
- Stem O.D.:17 mm
- Supplier No.:6220-75
- Top I.D.:75 mm
- Environmental certification:I-RECs, ISO 14001 - Monterrey
Specifications
About this item
PYREX®, borosilicate glass.
With short, wide stem. Used for transferring powders or filling bottles.
Customers Who Bought This Also Bought