PYREX® VISTA™ Round-Bottom Boiling Flasks, Corning
# de catalogue 89090-390
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:500 ml
- ST Joint No.:24/40
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:89090-390
- Supplier No.:70320-500
Spécifications
A propos de cet article
PYREX® 7740 borosilicate glass, clear, with short neck.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
These flasks have full length outer standard tapered joint. Wall thickness is controlled to provide balance between thermal expansion and mechanical strength for shock resistance.
Certifications: ASTM E-1403
Customers Who Bought This Also Bought