Pipette Tips, Sorenson BioScience
Catalog # 89082-390
Supplier: Sorenson Bioscience
Specifications
- Volume:50 - 1250 µl
- Tip Color:Natural
- Cat. no.:89082-390
- Tip Style:QuickRack, extended length
- Supplier No.:34770
Specifications
About this item
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Customers Who Bought This Also Bought