Barrier Pipette Tips, Sorenson BioScience
# de catalogue 89082-370
Fournisseur: Sorenson Bioscience
Spécifications
- Volume:1 - 300 µl
- Tip Color:Natural
- Filter:+
- Sterile:+
- Tip Style:Barrier tip, 300XT, low binding
- Supplier No.:30510T-X
Spécifications
A propos de cet article
The barrier pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment and feature an inert hydrophobic aerosol-blocking barrier.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips minimize sample retention.
Customers Who Bought This Also Bought