ContactUsLinkComponent
Pipette Tips
Barrier Pipette Tips, Sorenson BioScience
# de catalogue 89082-370
Fournisseur:  Sorenson Bioscience
Barrier Pipette Tips, Sorenson BioScience
# de catalogue 89082-370
Fournisseur:  Sorenson Bioscience
Spécifications
  • Volume:
    1 - 300 µl
  • Tip Color:
    Natural
  • Filter:
    +
  • Sterile:
    +
  • Tip Style:
    Barrier tip, 300XT, low binding
  • Supplier No.:
    30510T-X
Spécifications
A propos de cet article
The barrier pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment and feature an inert hydrophobic aerosol-blocking barrier.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips minimize sample retention.
Customers Who Bought This Also Bought