ContactUsLinkComponent
E.dek 10ul Tips tips
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 89082-354
Fournisseur:  Sorenson Bioscience
E.dek 10ul Tips tips
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 89082-354
Fournisseur:  Sorenson Bioscience
Spécifications
  • Volume:
    1 - 20 µL
  • Packed:
    e•dek™ Environmental Paperboard Racks
  • Cat. no.:
    89082-354
  • Filter:
    +
  • Tip Style:
    Graduated (1 µL, 5 µL, 10 µL)
  • Supplier No.:
    10400
Spécifications
A propos de cet article
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
  • Dual-shot tip design
  • Easy-to-seal
  • Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.
Customers Who Bought This Also Bought