OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 89082-354
Fournisseur: Sorenson Bioscience
Spécifications
- Volume:1 - 20 µL
- Packed:e•dek™ Environmental Paperboard Racks
- Cat. no.:89082-354
- Filter:+
- Tip Style:Graduated (1 µL, 5 µL, 10 µL)
- Supplier No.:10400
Spécifications
A propos de cet article
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
- Dual-shot tip design
- Easy-to-seal
- Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.
Customers Who Bought This Also Bought