ContactUsLinkComponent
TIP FILTER ONETOUCH 1000 PK768
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
Catalog # 89082-350
TIP FILTER ONETOUCH 1000 PK768
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
Catalog # 89082-350
Specifications
  • Volume:
    50 - 1250 µl
  • Packed:
    Plastic Racks
  • Cat. no.:
    89082-350
  • Sterile:
    +
  • Tip Style:
    Graduated (200 µL, 500 µL, 1000 µL)
  • Supplier No.:
    10380
Specifications
About this item
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
  • Dual-shot tip design
  • Easy-to-seal
  • Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.
Customers Who Bought This Also Bought