OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
Catalog # 89082-350
Supplier: Sorenson Bioscience
Specifications
- Volume:50 - 1250 µl
- Packed:Plastic Racks
- Cat. no.:89082-350
- Sterile:+
- Tip Style:Graduated (200 µL, 500 µL, 1000 µL)
- Supplier No.:10380
Specifications
About this item
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
- Dual-shot tip design
- Easy-to-seal
- Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.
Customers Who Bought This Also Bought