ART® Barrier Ultra Micro Tips with MicroPoint Design, Thermo Scientific
# de catalogue 89082-090
Fournisseur: Thermo Scientific
Spécifications
- Length:8.89 cm (31/2")
- Volume:1000 µl
- Sterile:+
- Tip Style:MicroPoint, Hinged Rack
- Supplier No.:2179-HR
Spécifications
A propos de cet article
Molecular Bioproducts ART® pipette tips are pre-sterilized and precision molded with the ART® self-sealing barrier.
They are guaranteed to prevent carryover contamination during aspiration and delivery of samples. Ideally suited for low volume applications in genetic studies, forensics, PCR, and radioisotope sampling.
Customers Who Bought This Also Bought