ContactUsLinkComponent
PCR Tube Caps
Axygen® PCR Tube Strips and Caps, Corning
# de catalogue 89080-526
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
PCR Tube Caps
Axygen® PCR Tube Strips and Caps, Corning
# de catalogue 89080-526
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Spécifications
  • Capacity:
    0.2 ml
  • Color:
    Clear
  • Description:
    8-Strip PCR flat caps
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Cat. no.:
    89080-526
  • Supplier No.:
    PCR-02-FCP-C
Spécifications
A propos de cet article
Axygen® polypropylene 8/12-tube thin wall PCR tube strips and caps are specifically designed for PCR use and are ideal for thermal transfer applications. The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling. Made from highly polished moulds and free of lubricants, dyes, heavy metals, and fillers. Certified RNase, DNase, and pyrogen free to ensure the purity of your reaction.
  • Ultra-thin walls of uniform thickness optimise heat transfer
  • Tongue at the end of each strip for ease of handling
  • Certified free from DNase, RNase and endotoxin
The polypropylene 8-strip tubes with individually attached caps come with added benefits that answer many customer needs. In addition to many of the features of a PCR tube strip, these feature a stronger connection between each tube and the caps attached. This minimizes the risk of contamination and provides improved ease for customers when handling.
Customers Who Bought This Also Bought