Axygen® Automation Tips for Tecan®Corning
Catalog # 89080-500
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:50 µl
- Packed:96/Rack, 10 Racks/Pack
- Cat. no.:89080-500
- Sterile:+
- Supplier No.:EV-50-R-S
Specifications
About this item
PP, with or without filter, sterile or non sterile.
- Black tips are conductive for liquid level-sensing protocols
- Optimised fine point for accurate dispensing
- Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Packaging: Packaged in plastic blister packs.
Customers Who Bought This Also Bought