ContactUsLinkComponent
VWR Signature* Electroporation Cuvette
VWR® Signature™ Disposable Electroporation Cuvettes
Catalog # 89047-210
Supplier:  VWR International
undefined
VWR® Signature™ Disposable Electroporation Cuvettes
Catalog # 89047-210
Supplier:  VWR International
Specifications
  • Volume:
    800 µl
  • Description:
    Round Cap
  • Cat. no.:
    89047-210
  • Gap Size:
    4 mm
Specifications
About this item
VWR Signature™ Disposable Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells.
  • Cuvettes are precision built to provide exacting tolerances, ensuring reproducible results
  • Polished aluminum plates reduce static and arcing
  • Compatible with most electroporation devices
  • Gamma irradiated for sterility
Disposable cuvettes are manufactured from high quality, durable polycarbonate material that has been tested to optimize transformation efficiencies.
Ordering information: Each pack contains 50 individually wrapped cuvettes.
Frequently Bought Together