Order Entry
ContactUsLinkComponent
VWR Buchner Filter Funnel
VWR® Buchner Filter Funnels
# de catalogue 89038-120
Fournisseur:  VWR International
VWR Buchner Filter Funnel
VWR® Buchner Filter Funnels
# de catalogue 89038-120
Fournisseur:  VWR International

Spécifications

  • Capacity:
    87 ml
  • Height:
    101 mm
  • Cat. no.:
    89038-120
  • Fits Paper Diameter:
    50-55 mm

Spécifications

A propos de cet article

These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.

  • Highly resistant to chemicals
  • Autoclavable
  • Glazed inside and out, except for the rim
Customers Who Bought This Also Bought