ContactUsLinkComponent
VWR Dessicator Plate
VWR® Desiccator Plates
# de catalogue 89038-072
Fournisseur:  VWR International
VWR Dessicator Plate
VWR® Desiccator Plates
# de catalogue 89038-072
Fournisseur:  VWR International
Spécifications
  • Ø:
    230 mm
  • Hole Diameter:
    5 mm
  • Cat. no.:
    89038-072
Spécifications
A propos de cet article
Chemical-porcelain dessicator plates are perforated with numerous small holes and a large center hole.
  • Autoclavable
  • Plates without feet
Plates are also glazed on one side and highly resistant to chemicals. For use in temperatures up to 1150 °C (2102 °F).
Customers Who Bought This Also Bought