VWR® Desiccator Plates
# de catalogue 89038-070
Fournisseur: VWR International
Spécifications
- Ø:190 mm
- Hole Diameter:5 mm
- Cat. no.:89038-070
Spécifications
A propos de cet article
Chemical-porcelain dessicator plates are perforated with numerous small holes and a large center hole.
- Autoclavable
- Plates without feet
Plates are also glazed on one side and highly resistant to chemicals. For use in temperatures up to 1150 °C (2102 °F).
Customers Who Bought This Also Bought