Order Entry
ContactUsLinkComponent
VWR Porous Bottom Crucible
VWR® Porous-Bottom Crucibles, Porcelain
# de catalogue 89038-050
Fournisseur:  VWR International
VWR Porous Bottom Crucible
VWR® Porous-Bottom Crucibles, Porcelain
# de catalogue 89038-050
Fournisseur:  VWR International

Spécifications

  • Capacity:
    25 ml
  • Cat. no.:
    89038-050
  • O.D.×H:
    36×42 mm
  • Pore Diameter:
    15 µm

Spécifications

A propos de cet article

Chemical-porcelain crucibles feature a porous bottom with a porous disc permanently formed into the walls.

Cover is ordered separately.

  • Stable porosity
  • Consistent rate of flow
  • Chemical-porcelain construction
  • Cover is ordered separately.

Chemical-porcelain crucibles feature a porous bottom with a porous disc permanently formed into the walls.

Customers Who Bought This Also Bought