Order Entry
ContactUsLinkComponent
VWR Crucibles Low Form
VWR® High and Low Form Porcelain Crucibles
Catalog # 89038-026
Supplier:  VWR International
VWR Crucibles Low Form
VWR® High and Low Form Porcelain Crucibles
Catalog # 89038-026
Supplier:  VWR International

Specifications

  • Ø:
    50 mm
  • Capacity:
    30 ml
  • Height:
    31 mm
  • Cat. no.:
    89038-026

Specifications

About this item

Chemical-porcelain crucibles are glazed inside and out, except the outside bottom and rim.

  • Glazed interior and exterior
  • Crucible covers are completely glazed, except for the rim
  • For use in temperatures up to 1150 °C (2102 °F)
Customers Who Bought This Also Bought