Order Entry
Canada
ContactUsLinkComponent
VWR® High and Low Form Porcelain Crucibles
Catalogue # 89037-996
Fournisseur:  VWR International
undefined
VWR® High and Low Form Porcelain Crucibles
Catalogue # 89037-996
Fournisseur:  VWR International

Spécifications

  • Ø:
    50 mm
  • Capacity:
    40 ml
  • Height:
    40 mm
  • Cat. no.:
    89037-996

Spécifications

À propos de cet article

Chemical-porcelain crucibles are glazed inside and out, except the outside bottom and rim.

  • Glazed interior and exterior
  • Crucible covers are completely glazed, except for the rim
  • For use in temperatures up to 1150 °C (2102 °F)
Frequently Bought Together