VWR® Mortars and Pestles, Agate
# de catalogue 89037-502
Fournisseur: VWR International
Spécifications
- Length:51 mm
- Description:for Mortar 89037-484
- Cat. no.:89037-502
Spécifications
A propos de cet article
Mortars and pestles are made of thick-walled agate, which is the most common variety of chalcedony, a semiprecious, very hard silica mineral.
- Thick-walled agate construction
- Highly polished surfaces
- Octagonal shape
They feature highly polished, glossy grinding surfaces. Mortar is octagonal in shape.
Ordering information: Supplied in mortar and pestle sets or pestles only.
Customers Who Bought This Also Bought