ContactUsLinkComponent
VWR Pestle Only Agate
VWR® Mortars and Pestles, Agate
# de catalogue 89037-502
Fournisseur:  VWR International
VWR Pestle Only Agate
VWR® Mortars and Pestles, Agate
# de catalogue 89037-502
Fournisseur:  VWR International
Spécifications
  • Length:
    51 mm
  • Description:
    for Mortar 89037-484
  • Cat. no.:
    89037-502
Spécifications
A propos de cet article
Mortars and pestles are made of thick-walled agate, which is the most common variety of chalcedony, a semiprecious, very hard silica mineral.
  • Thick-walled agate construction
  • Highly polished surfaces
  • Octagonal shape
They feature highly polished, glossy grinding surfaces. Mortar is octagonal in shape.
Ordering information: Supplied in mortar and pestle sets or pestles only.
Customers Who Bought This Also Bought