VWR® Mortars and Pestles, Agate
# de catalogue 89037-480
Fournisseur: VWR International
Spécifications
- Height:19 mm
- Top O.D.:25 mm
- Top I.D.:20 mm
Spécifications
A propos de cet article
Mortars and pestles are made of thick-walled agate, which is the most common variety of chalcedony, a semiprecious, very hard silica mineral.
- Thick-walled agate construction
- Highly polished surfaces
- Octagonal shape
They feature highly polished, glossy grinding surfaces. Mortar is octagonal in shape.
Ordering information: Supplied in mortar and pestle sets or pestles only.
Customers Who Bought This Also Bought