ContactUsLinkComponent
Pipet Tips, 300ul Clear
ART® and Hydrologix (HLT) SoftFit-L™ Pipette Tips for Rainin® LTS Pipettors, Molecular BioProducts
# de catalogue 89031-400
Fournisseur:  Thermo Scientific
ART® and Hydrologix (HLT) SoftFit-L™ Pipette Tips for Rainin® LTS Pipettors, Molecular BioProducts
# de catalogue 89031-400
Fournisseur:  Thermo Scientific
Spécifications
  • Volume:
    300 µl
  • Packed:
    Hinged Rack
  • Tip Style:
    HLT
  • Supplier No.:
    3732-HR
Spécifications
A propos de cet article
SoftFit-L™ pipette tips are available in five volume sizes, and are designed for use with Rainin® LTS pipettors.
  • Certified pure
  • Reload inserts are color-coded by size
Tips deliver highly accurate samples, seal well on the pipettor, and are easy to seat and eject. To accommodate a variety of applications, SoftFit-L™ pipette tips offer an expanded selection of ART®, HLT, and low retention designs. The ART® (Aerosol Resistant Tips) series features a self-sealing barrier designed to prevent contamination even under the most demanding conditions.
These tips guarantee 100% protection against aerosol and liquid contamination. Low retention tips are designed to reduce sample retention. Molded from virgin polypropylene, they do not use additives or coatings that may affect sample integrity. Low retention tips ensure reduced sample loss due to adhesion, allowing for more accurate dispensing and an increased level of reproducibility from one procedure to the next.
Customers Who Bought This Also Bought