Axygen® Pipette Tip Reload System, Corning
# de catalogue 89029-919
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:1 - 200 µl
- Tip Color:Clear
- Tip Style:Universal Fit
- Supplier No.:RFL-222-C
Spécifications
A propos de cet article
Gilson®-style and universal fit pipette tips in easy-to-load refill racks.
Gilson®-style and universal fit pipet tips in easy-to-load refill racks. Universal fit tips have a beveled reference and graduated marks.
- RNase-/DNase-free
- Nonpyrogenic
- Translucent rack allows tips to be easily seen
- Color-coded insert allows for easy identification of tip
Universal fit tips have a beveled reference and graduated marks.
Axygen® tip reload system contains durable refill rack that can be easily refilled by hand and is autoclavable. They are available in different colors and are packaged in bulk or racks.
Customers Who Bought This Also Bought