ContactUsLinkComponent
Viton* O-Ring #331
Viton® O-Rings, Chemglass
# de catalogue 89029-276
Fournisseur:  Chemglass
Small Business Enterprise
Viton* O-Ring #331
Viton® O-Rings, Chemglass
# de catalogue 89029-276
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Cat. no.:
    89029-276
  • Supplier No.:
    CG-305-331
Spécifications
A propos de cet article
Brown, low compression, 75 durometer Viton® O-rings are suitable for acids, aliphatic hydrocarbons, oxidizing agents, and all petroleum products
Note: Viton® O-ring 89029-223 is not a standard size.
They are especially good for vacuum service and low gas permeability. Temperature range: –30 to 200°C (–22 to 392°F).
Customers Who Bought This Also Bought